Product Details

SNP ID
rs12121124
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:58576329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGACCACCACGATGACGGCGATGAG[A/G]CCGGCGGTGAGGCGCTTCATGGAGA
Phenotype
MIM: 137290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TACSTD2 PubMed Links

Gene Details

Gene
TACSTD2
Gene Name
tumor-associated calcium signal transducer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002353.2 1166 Silent Mutation GGC,GGT G276G NP_002344.2

View Full Product Details