Product Details
- SNP ID
-
rs34856868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:92088726 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTAAGGGAAGAATTCCATACAGAT[A/G]TTACCTTCTCTGTGGGTTGTACTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BTBD8
PubMed Links
Gene Details
- Gene
- BTBD8
- Gene Name
- BTB domain containing 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183242.3 |
445 |
Missense Mutation |
ATT,GTT |
I60V |
NP_899065.2 |
View Full Product Details