Product Details
- SNP ID
-
rs16990421
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:32415897 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAGAAATGGGTCAGTTAAGAGG[C/T]GAGATTTGCATTCTTACCTCAAGAA
- Phenotype
-
MIM: 614109
MIM: 613901
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BPIFC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115332147] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BPIFC
- Gene Name
- BPI fold containing family C
- Gene
- RTCB
- Gene Name
- RNA 2',3'-cyclic phosphate and 5'-OH ligase
There are no transcripts associated with this gene.
View Full Product Details