Product Details

SNP ID
rs78663425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49487735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCTCTGTCTTGCATGTTTTGC[G/T]GAGCTTAACATCCATAATGAAGCAA
Phenotype
MIM: 600007 MIM: 180471
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FLT3LG PubMed Links

Gene Details

Gene
FLT3LG
Gene Name
fms related tyrosine kinase 3 ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204502.1 Intron NP_001191431.1
NM_001204503.1 Intron NP_001191432.1
NM_001278637.1 Intron NP_001265566.1
NM_001278638.1 Intron NP_001265567.1
NM_001459.3 Intron NP_001450.2
XM_005258680.4 Intron XP_005258737.3
XM_005258681.4 Intron XP_005258738.3
XM_005258682.4 Intron XP_005258739.3
XM_005258683.4 Intron XP_005258740.3
XM_006723116.3 Intron XP_006723179.2
XM_011526675.2 Intron XP_011524977.1
XM_011526676.2 Intron XP_011524978.1
XM_011526677.2 Intron XP_011524979.1
XM_011526678.2 Intron XP_011524980.1
XM_011526680.2 Intron XP_011524982.1
XM_011526682.2 Intron XP_011524984.1
XM_017026532.1 Intron XP_016882021.1
XM_017026533.1 Intron XP_016882022.1
XM_017026534.1 Intron XP_016882023.1
XM_017026535.1 Intron XP_016882024.1
Gene
RPL13A
Gene Name
ribosomal protein L13a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270491.1 Intron NP_001257420.1
NM_012423.3 Intron NP_036555.1
Gene
RPS11
Gene Name
ribosomal protein S11
There are no transcripts associated with this gene.

Gene
SNORD32A
Gene Name
small nucleolar RNA, C/D box 32A
There are no transcripts associated with this gene.

Gene
SNORD33
Gene Name
small nucleolar RNA, C/D box 33
There are no transcripts associated with this gene.

Gene
SNORD34
Gene Name
small nucleolar RNA, C/D box 34
There are no transcripts associated with this gene.

Gene
SNORD35A
Gene Name
small nucleolar RNA, C/D box 35A
There are no transcripts associated with this gene.

Gene
SNORD35B
Gene Name
small nucleolar RNA, C/D box 35B
There are no transcripts associated with this gene.

View Full Product Details