Product Details

SNP ID
rs192902107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199955776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTCCATACAGCACCGGCAGG[A/C]ACCGGTGTGAAGGGTCATAAAAATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf47 PubMed Links

Gene Details

Gene
C2orf47
Gene Name
chromosome 2 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024520.2 211 UTR 5 NP_078796.2
XM_005246852.2 211 UTR 5 XP_005246909.1
Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039693.2 211 Intron NP_001034782.1
XM_017003310.1 211 Intron XP_016858799.1

View Full Product Details