Product Details

SNP ID
rs17094161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73950133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCCCTTTCTAGCTTTGGCGTCT[A/G]GTTGGCTTCCAGGGCAGCCTCCGAT
Phenotype
MIM: 614647 MIM: 603162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ6 PubMed Links

Gene Details

Gene
COQ6
Gene Name
coenzyme Q6, monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182476.2 200 Intron NP_872282.1
NM_182480.2 200 Nonsense Mutation TAG,TGG *14W NP_872286.2
XM_006720156.1 200 Intron XP_006720219.1
XM_011536807.1 200 Intron XP_011535109.1
XM_011536808.1 200 Intron XP_011535110.1
XM_011536809.2 200 UTR 5 XP_011535111.1
XM_011536810.2 200 Intron XP_011535112.1
XM_017021351.1 200 Intron XP_016876840.1
XM_017021352.1 200 Intron XP_016876841.1
Gene
ENTPD5
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
There are no transcripts associated with this gene.

Gene
FAM161B
Gene Name
family with sequence similarity 161 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152445.2 200 Missense Mutation CCA,CTA P28L NP_689658.2
XM_011536475.1 200 Missense Mutation CCA,CTA P28L XP_011534777.1

View Full Product Details