Product Details

SNP ID
rs12006467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35090723 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCACTTCTTATTCCCTAATCCA[C/T]AGGCTACTGCTCCACAATCATATAC
Phenotype
MIM: 614730 MIM: 608292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGO PubMed Links

Gene Details

Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201484.1 Intron NP_001188413.1
NM_032634.3 Intron NP_116023.2
NM_152850.3 Intron NP_690577.2
XM_005251619.3 Intron XP_005251676.1
XM_017015222.1 Intron XP_016870711.1
XM_017015223.1 Intron XP_016870712.1
XM_017015224.1 Intron XP_016870713.1
Gene
STOML2
Gene Name
stomatin like 2
There are no transcripts associated with this gene.

View Full Product Details