Product Details

SNP ID
rs61741273
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88180268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATGAAATCTCCCTCCCCCCACA[C/T]TATTTTCTGTAAAATGCCTTTTTTT
Phenotype
MIM: 605873
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNK10 PubMed Links

Gene Details

Gene
KCNK10
Gene Name
potassium two pore domain channel subfamily K member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021161.4 7350 UTR 3 NP_066984.1
NM_138317.2 7350 UTR 3 NP_612190.1
NM_138318.2 7350 UTR 3 NP_612191.1
XM_011536840.2 7350 UTR 3 XP_011535142.1
Gene
LOC105370612
Gene Name
uncharacterized LOC105370612
There are no transcripts associated with this gene.

View Full Product Details