Product Details

SNP ID
rs116478535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31887179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACAAAACCAGTTACATCCTTT[C/T]CCTTTCCTCATTGCAGAACCAAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTLL9 PubMed Links

Gene Details

Gene
TTLL9
Gene Name
tubulin tyrosine ligase like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008409.2 Intron NP_001008409.1
XM_005260309.2 Intron XP_005260366.1
XM_006723723.2 Intron XP_006723786.1
XM_006723724.2 Intron XP_006723787.1
XM_011528617.1 Intron XP_011526919.1
XM_011528618.1 Intron XP_011526920.1
XM_011528619.1 Intron XP_011526921.1
XM_011528620.2 Intron XP_011526922.1
XM_011528621.1 Intron XP_011526923.1
XM_011528622.1 Intron XP_011526924.1
XM_011528623.2 Intron XP_011526925.1
XM_011528624.1 Intron XP_011526926.1
XM_011528625.1 Intron XP_011526927.1
XM_011528626.1 Intron XP_011526928.1
XM_011528627.2 Intron XP_011526929.1
XM_011528628.1 Intron XP_011526930.1
XM_011528629.1 Intron XP_011526931.1
XM_011528630.1 Intron XP_011526932.1
XM_011528631.1 Intron XP_011526933.1
XM_011528632.1 Intron XP_011526934.1
XM_011528633.1 Intron XP_011526935.1
XM_011528634.1 Intron XP_011526936.1
XM_011528635.1 Intron XP_011526937.1
XM_011528637.2 Intron XP_011526939.1
XM_011528638.1 Intron XP_011526940.1
XM_011528639.1 Intron XP_011526941.1
XM_011528640.1 Intron XP_011526942.1
XM_011528641.1 Intron XP_011526943.1
XM_011528643.2 Intron XP_011526945.1
XM_011528644.1 Intron XP_011526946.1
XM_011528646.1 Intron XP_011526948.1
XM_017027694.1 Intron XP_016883183.1
XM_017027695.1 Intron XP_016883184.1
XM_017027696.1 Intron XP_016883185.1
XM_017027697.1 Intron XP_016883186.1
XM_017027698.1 Intron XP_016883187.1
XM_017027699.1 Intron XP_016883188.1
XM_017027700.1 Intron XP_016883189.1
XM_017027701.1 Intron XP_016883190.1
XM_017027702.1 Intron XP_016883191.1
XM_017027703.1 Intron XP_016883192.1

View Full Product Details