Product Details

SNP ID
rs76836760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:133052376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGCCGTAAGTGTGGGCCGGGCC[G/T]GCCGCGTGCACTGGGGTTAGGAGAT
Phenotype
MIM: 601113
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPA4 PubMed Links

Gene Details

Gene
HSPA4
Gene Name
heat shock protein family A (Hsp70) member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002154.3 Intron NP_002145.3

View Full Product Details