Product Details

SNP ID
rs879740307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74732499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATATCATTTTACATATTTCCA[A/T]ATTTTCCCCCCATTTCCTAACCTAT
Phenotype
MIM: 607009
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRPM6 PubMed Links

Gene Details

Gene
TRPM6
Gene Name
transient receptor potential cation channel subfamily M member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177310.1 Intron NP_001170781.1
NM_001177311.1 Intron NP_001170782.1
NM_017662.4 Intron NP_060132.3

View Full Product Details