Product Details
- SNP ID
-
rs61741101
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:111414593 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGAACAAAGCGGTTGTCAAAAGC[C/T]AGAGGAGTTTGTTCCGGGAGCTGGA
- Phenotype
-
MIM: 603578
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OVGP1
PubMed Links
Gene Details
- Gene
- OVGP1
- Gene Name
- oviductal glycoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002557.3 |
1964 |
Silent Mutation |
CTA,CTG |
L636L |
NP_002548.3 |
View Full Product Details