Product Details

SNP ID
rs77039362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49002747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAATCCATTCTCTTTCCTCCCC[C/G]ATACCTTCCCTAGCTCCCAGATAGA
Phenotype
MIM: 610588 MIM: 602742
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDN PubMed Links

Gene Details

Gene
DDN
Gene Name
dendrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015086.1 1254 Intron NP_055901.2
XM_011538055.2 1254 Intron XP_011536357.1
Gene
PRKAG1
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206709.1 1254 UTR 3 NP_001193638.1
NM_001206710.1 1254 UTR 3 NP_001193639.1
NM_002733.4 1254 UTR 3 NP_002724.1
XM_005269019.4 1254 UTR 3 XP_005269076.1
XM_005269020.2 1254 UTR 3 XP_005269077.1
XM_006719499.2 1254 UTR 3 XP_006719562.1
XM_011538562.2 1254 UTR 3 XP_011536864.1
XM_017019622.1 1254 UTR 3 XP_016875111.1
XM_017019623.1 1254 UTR 3 XP_016875112.1

View Full Product Details