Product Details

SNP ID
rs35068602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71120055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGAAGAGGTGTTTGGGTACTT[A/G]GCCAAATACACAGTGCCTGTGATGC
Phenotype
MIM: 308380 MIM: 300188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL2RG PubMed Links

Gene Details

Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
There are no transcripts associated with this gene.

Gene
MED12
Gene Name
mediator complex subunit 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005120.2 637 Silent Mutation TTA,TTG L146L NP_005111.2

View Full Product Details