Product Details
- SNP ID
-
hCV25958477
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132815206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGTTCTGAATGGCATCCGTCCT[A/C]CCGGAAACCTAGGAAAAGGTATCAG
- Phenotype
-
MIM: 603660
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RPS12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74627356] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPS12
- Gene Name
- ribosomal protein S12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001016.3 |
|
Intron |
|
|
NP_001007.2 |
- Gene
- SNORA33
- Gene Name
- small nucleolar RNA, H/ACA box 33
There are no transcripts associated with this gene.
- Gene
- SNORD100
- Gene Name
- small nucleolar RNA, C/D box 100
There are no transcripts associated with this gene.
- Gene
- SNORD101
- Gene Name
- small nucleolar RNA, C/D box 101
There are no transcripts associated with this gene.
View Full Product Details