Product Details
- SNP ID
-
rs113556292
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40750352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGCAAGCAGATTTCAGTATATTA[C/T]GCCATCTATGCAGATTATTTTACCT
- Phenotype
-
MIM: 616646
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT25
PubMed Links
Gene Details
- Gene
- KRT25
- Gene Name
- keratin 25
View Full Product Details