Product Details

SNP ID
rs113742037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7310526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTCTGTTCAGTGCAGGGCCCT[C/T]TGTCCTTCCCAGGGATCATCTTCAT
Phenotype
MIM: 614360 MIM: 606079
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM4 PubMed Links

Gene Details

Gene
ACSM4
Gene Name
acyl-CoA synthetase medium-chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080454.1 Intron NP_001073923.1
Gene
CD163L1
Gene Name
CD163 molecule like 1
There are no transcripts associated with this gene.

View Full Product Details