Product Details

SNP ID
rs11548606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67508194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCTGCACGCCCCTGGTAAATAG[A/C]GGTCTGGCCGGAAGGGCGAGCGGGC
Phenotype
MIM: 608794
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDK2AP2 PubMed Links

Gene Details

Gene
CDK2AP2
Gene Name
cyclin dependent kinase 2 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271849.1 535 Intron NP_001258778.1
NM_005851.4 535 UTR 5 NP_005842.1
Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
There are no transcripts associated with this gene.

View Full Product Details