Product Details

SNP ID
rs45591543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:89576997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATCGCTCTAATCATTGAAAAG[C/T]GCCCTGTGCTGAGTAAAGCAGCCAG
Phenotype
MIM: 602628
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXN3 PubMed Links

Gene Details

Gene
FOXN3
Gene Name
forkhead box N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085471.1 Intron NP_001078940.1
NM_005197.3 Intron NP_005188.2
Gene
FOXN3-AS2
Gene Name
FOXN3 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details