Product Details

SNP ID
rs7200194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53495378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGGATACAAATGACACATGCA[C/G]ATCAAACCACCCCACATTCACTTTG
Phenotype
MIM: 608483 MIM: 180203
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AKTIP PubMed Links

Gene Details

Gene
AKTIP
Gene Name
AKT interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012398.2 Intron NP_001012398.1
NM_001308325.1 Intron NP_001295254.1
NM_022476.3 Intron NP_071921.1
XM_005256095.4 Intron XP_005256152.1
XM_005256096.4 Intron XP_005256153.1
XM_005256097.4 Intron XP_005256154.1
XM_005256098.4 Intron XP_005256155.1
XM_017023564.1 Intron XP_016879053.1
XM_017023565.1 Intron XP_016879054.1
XM_017023566.1 Intron XP_016879055.1
Gene
RBL2
Gene Name
RB transcriptional corepressor like 2
There are no transcripts associated with this gene.

View Full Product Details