Product Details

SNP ID
hCV25970837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:139730949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGAGGGCTGGGAGGGGTCTGG[A/G]GGAGGAGGAGGAGATGGGGCTGCAG
Phenotype
MIM: 611966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC9 PubMed Links

Gene Details

Gene
TRAPPC9
Gene Name
trafficking protein particle complex 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160372.3 3666 UTR 3 NP_001153844.1
NM_001321646.1 3666 UTR 3 NP_001308575.1
NM_031466.7 3666 UTR 3 NP_113654.4

View Full Product Details