Product Details

SNP ID
rs112745062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26584185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCCGCGGCTTGGCGGGCTAGG[A/G]CAGGGGAAATGTTGCAGGAGGAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D19 PubMed Links

Gene Details

Gene
TBC1D19
Gene Name
TBC1 domain family member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292054.1 102 UTR 5 NP_001278983.1
NM_018317.3 102 UTR 5 NP_060787.2
XM_011513852.2 102 UTR 5 XP_011512154.1
XM_017008363.1 102 UTR 5 XP_016863852.1
XM_017008364.1 102 UTR 5 XP_016863853.1

View Full Product Details