Product Details

SNP ID
rs35286445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70518820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTTTTGTCTTCTAGGATCAACT[C/T]GGTCATTACCAGTAAGTATGTTATG
Phenotype
MIM: 610912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMTN PubMed Links

Gene Details

Gene
AMTN
Gene Name
amelotin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286731.1 141 Missense Mutation CGG,TGG R15W NP_001273660.1
NM_212557.3 141 Missense Mutation CGG,TGG R15W NP_997722.1

View Full Product Details