Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_005259582.4 | Intron | XP_005259639.1 | |||
XM_006722769.3 | Intron | XP_006722832.1 | |||
XM_011528072.2 | Intron | XP_011526374.1 | |||
XM_011528073.1 | Intron | XP_011526375.1 | |||
XM_011528074.2 | Intron | XP_011526376.1 | |||
XM_011528075.2 | Intron | XP_011526377.1 | |||
XM_011528077.2 | Intron | XP_011526379.1 | |||
XM_011528081.2 | Intron | XP_011526383.1 | |||
XM_011528082.2 | Intron | XP_011526384.1 |