Product Details

SNP ID
rs17094164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73950234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCGCATTTTATTTCCGGTTCTGAG[C/G]ACGCCGCGGAAGCGGGACGGGATTG
Phenotype
MIM: 614647 MIM: 603162
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COQ6 PubMed Links

Gene Details

Gene
COQ6
Gene Name
coenzyme Q6, monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182476.2 181 Intron NP_872282.1
NM_182480.2 181 Intron NP_872286.2
XM_006720156.1 181 Intron XP_006720219.1
XM_011536807.1 181 Intron XP_011535109.1
XM_011536808.1 181 Intron XP_011535110.1
XM_011536809.2 181 Intron XP_011535111.1
XM_011536810.2 181 Intron XP_011535112.1
XM_017021351.1 181 Intron XP_016876840.1
XM_017021352.1 181 Intron XP_016876841.1
Gene
ENTPD5
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
There are no transcripts associated with this gene.

Gene
FAM161B
Gene Name
family with sequence similarity 161 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152445.2 181 UTR 5 NP_689658.2
XM_011536475.1 181 Intron XP_011534777.1

View Full Product Details