Product Details

SNP ID
rs10856852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:84976382 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGATACTTAAGAGCCAGGATATC[A/G]TCTGCCATGCTTTTTTTGCCCTATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDFY3 PubMed Links

Gene Details

Gene
WDFY3
Gene Name
WD repeat and FYVE domain containing 3
There are no transcripts associated with this gene.

Gene
WDFY3-AS2
Gene Name
WDFY3 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details