Product Details

SNP ID
rs2233166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87504836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCCGCCGGCCGGAAGCGCCCT[C/G]CAGACCCGCCCCCTGCCCTCCGGAG
Phenotype
MIM: 605391
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MINPP1 PubMed Links

Gene Details

Gene
MINPP1
Gene Name
multiple inositol-polyphosphate phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178117.1 2045 UTR 5 NP_001171588.1
NM_001178118.1 2045 Intron NP_001171589.1
NM_004897.4 2045 UTR 5 NP_004888.2
XM_006718078.2 2045 UTR 5 XP_006718141.1
XM_011540379.2 2045 Intron XP_011538681.1
XM_017016965.1 2045 UTR 5 XP_016872454.1
XM_017016966.1 2045 Intron XP_016872455.1
Gene
MIR4678
Gene Name
microRNA 4678
There are no transcripts associated with this gene.

View Full Product Details