Product Details

SNP ID
rs2233164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87504719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTCCGCCGGCTCCACCCGAGCC[A/T]CAAGTCTACAGGACTCCTCGGAGCT
Phenotype
MIM: 605391
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MINPP1 PubMed Links

Gene Details

Gene
MINPP1
Gene Name
multiple inositol-polyphosphate phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178117.1 1928 UTR 5 NP_001171588.1
NM_001178118.1 1928 Intron NP_001171589.1
NM_004897.4 1928 UTR 5 NP_004888.2
XM_006718078.2 1928 UTR 5 XP_006718141.1
XM_011540379.2 1928 Intron XP_011538681.1
XM_017016965.1 1928 UTR 5 XP_016872454.1
XM_017016966.1 1928 Intron XP_016872455.1
Gene
MIR4678
Gene Name
microRNA 4678
There are no transcripts associated with this gene.

View Full Product Details