Product Details

SNP ID
rs1127201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34978412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTTTAACCACCACCTACTTCC[A/C]TTAAACACACACACATTTTTATCTT
Phenotype
MIM: 610548
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AQR PubMed Links

Gene Details

Gene
AQR
Gene Name
aquarius intron-binding spliceosomal factor
There are no transcripts associated with this gene.

Gene
ZNF770
Gene Name
zinc finger protein 770
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014106.3 Intron NP_054825.2
XM_011521744.2 Intron XP_011520046.1

View Full Product Details