Product Details

SNP ID
rs13171770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160196946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCCACTCAGCCACTCTCATCC[C/T]GCTTTCCTACTCCAGACCTTATTAG
Phenotype
MIM: 600422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP6 PubMed Links

Gene Details

Gene
FABP6
Gene Name
fatty acid binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040442.1 Intron NP_001035532.1
NM_001130958.1 Intron NP_001124430.1
NM_001445.2 Intron NP_001436.1

View Full Product Details