Product Details

SNP ID
rs3749824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133080220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTTTGGGGGAATGTAGTCGCC[A/G]GAGCTCTGCGTTGTCTTCCGGCTTT
Phenotype
MIM: 604082
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300776.1 18 UTR 5 NP_001287705.1
NM_001300777.1 18 UTR 5 NP_001287706.1
NM_001300778.1 18 UTR 5 NP_001287707.1
NM_003440.3 18 UTR 5 NP_003431.2
XM_005266188.4 18 Intron XP_005266245.1
XM_011534833.2 18 Intron XP_011533135.1
XM_011534834.2 18 UTR 5 XP_011533136.1
XM_011534835.2 18 UTR 5 XP_011533137.1
XM_011534836.2 18 Intron XP_011533138.1
XM_011534837.2 18 Intron XP_011533139.1
XM_011534838.2 18 Intron XP_011533140.1
XM_011534839.2 18 Intron XP_011533141.1
XM_011534840.2 18 Intron XP_011533142.1
XM_017019924.1 18 Intron XP_016875413.1
XM_017019925.1 18 UTR 5 XP_016875414.1

View Full Product Details