Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243013.1 | 2540 | Intron | NP_001229942.1 | ||
NM_001243014.1 | 2540 | Intron | NP_001229943.1 | ||
NM_019624.3 | 2540 | Intron | NP_062570.1 | ||
NM_019625.3 | 2540 | Intron | NP_062571.1 | ||
NM_203444.3 | 2540 | UTR 3 | NP_982269.2 | ||
XM_005253558.2 | 2540 | Intron | XP_005253615.1 | ||
XM_011538095.2 | 2540 | Intron | XP_011536397.1 | ||
XM_011538096.2 | 2540 | Intron | XP_011536398.1 | ||
XM_011538098.2 | 2540 | Intron | XP_011536400.1 | ||
XM_011538099.2 | 2540 | Intron | XP_011536401.1 | ||
XM_017019103.1 | 2540 | Intron | XP_016874592.1 | ||
XM_017019104.1 | 2540 | Intron | XP_016874593.1 |