Product Details

SNP ID
rs8111858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4326242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTGTATCTGTGCTACATAGCAT[G/T]TGTATGTGACTGGAATTGGGGCACA
Phenotype
MIM: 609828 MIM: 607881
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FSD1 PubMed Links

Gene Details

Gene
FSD1
Gene Name
fibronectin type III and SPRY domain containing 1
There are no transcripts associated with this gene.

Gene
STAP2
Gene Name
signal transducing adaptor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013841.1 Intron NP_001013863.1
NM_017720.2 Intron NP_060190.2
XM_011528123.1 Intron XP_011526425.1

View Full Product Details