Product Details

SNP ID
rs2732614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46286843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATACAGTACAATTCCTCACTGCTG[T/C]CCTCTTGTGCCCATTTACAATCATT
Phenotype
MIM: 616555
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ARL17B PubMed Links

Gene Details

Gene
ARL17B
Gene Name
ADP ribosylation factor like GTPase 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039083.3 Intron NP_001034172.3
NM_001103154.1 Intron NP_001096624.1
XM_005256918.4 Intron XP_005256975.1
XM_011524160.2 Intron XP_011522462.1
XM_011524161.2 Intron XP_011522463.1
XM_011524162.2 Intron XP_011522464.1
XM_011524163.2 Intron XP_011522465.1
XM_011524164.2 Intron XP_011522466.1
XM_011524165.2 Intron XP_011522467.1
XM_011524166.2 Intron XP_011522468.1
XM_011524167.2 Intron XP_011522469.1
Gene
LRRC37A
Gene Name
leucine rich repeat containing 37A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014834.4 Intron NP_055649.4
XM_005257879.4 Intron XP_005257936.1
XM_005257880.4 Intron XP_005257937.1
XM_005257882.4 Intron XP_005257939.1
XM_005257883.4 Intron XP_005257940.1
XM_005257884.4 Intron XP_005257941.1
XM_006722210.3 Intron XP_006722273.1
XM_006722211.3 Intron XP_006722274.1
XM_011525539.2 Intron XP_011523841.1
XM_017025472.1 Intron XP_016880961.1
XM_017025473.1 Intron XP_016880962.1

View Full Product Details