Product Details

SNP ID
rs3750182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125431292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCCCGGGCCAGGACCCGGCTG[G/T]GGTCGGGGGCGCGGGACGGGGTGGT
Phenotype
MIM: 609461
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIB1 PubMed Links

Gene Details

Gene
TRIB1
Gene Name
tribbles pseudokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282985.1 Intron NP_001269914.1
NM_025195.3 Intron NP_079471.1

View Full Product Details