Product Details
- SNP ID
-
rs6793076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:191204208 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCAGCTGATATTTTAATTTTTTTT[A/T]AACGGGAAATGTTAAATTTCTCAAT
- Phenotype
-
MIM: 610280
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OSTN
PubMed Links
Gene Details
- Gene
- OSTN
- Gene Name
- osteocrin
- Gene
- OSTN-AS1
- Gene Name
- OSTN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details