Product Details

SNP ID
rs12823432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113425125 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGGGGAAACAGGCACAGAGAG[A/G]TTGTGACACTTGTTTGGGTCACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDSL PubMed Links

Gene Details

Gene
SDSL
Gene Name
serine dehydratase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304993.1 Intron NP_001291922.1
NM_138432.3 Intron NP_612441.1
XM_005253831.4 Intron XP_005253888.1
XM_011537846.2 Intron XP_011536148.1
XM_017018763.1 Intron XP_016874252.1

View Full Product Details