Product Details

SNP ID
rs12804219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94962630 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGATAACAAAACTGTTATTTCC[A/G]AAGGTTTTATTTTATATATTGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CWC15 PubMed Links

Gene Details

Gene
CWC15
Gene Name
CWC15 spliceosome-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016403.3 1568 UTR 3 NP_057487.2
XM_011542868.2 1568 Intron XP_011541170.1
XM_017017916.1 1568 Intron XP_016873405.1

View Full Product Details