Product Details

SNP ID
rs10802624
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:235331656 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTTTTTTTTAGAAGATTC[A/G]TAGCTTTCATCATATTCTCAAAAGG
Phenotype
MIM: 609696 MIM: 606982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID4B PubMed Links

Gene Details

Gene
ARID4B
Gene Name
AT-rich interaction domain 4B
There are no transcripts associated with this gene.

Gene
GGPS1
Gene Name
geranylgeranyl diphosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037277.1 Intron NP_001032354.1

View Full Product Details