Product Details

SNP ID
rs2016257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:126663355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTCCTTCATGTTTTTATCCA[G/A]GAGTCAGCACTGCGGTGGAGACAGC
Phenotype
MIM: 110750
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GYPC PubMed Links

Gene Details

Gene
GYPC
Gene Name
glycophorin C (Gerbich blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256584.1 Intron NP_001243513.1
NM_002101.4 Intron NP_002092.1
NM_016815.3 Intron NP_058131.1
XM_006712460.3 Intron XP_006712523.1

View Full Product Details