Product Details
- SNP ID
-
rs7223035
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:6868225 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATACAATAGTATCTTTAACTGGGGG[A/G]AAAAAAAGAAGGAGGAGAAGAAGAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALOX12P2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147796267] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALOX12P2
- Gene Name
- arachidonate 12-lipoxygenase pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details