Product Details

SNP ID
rs1061955
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:149942458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGAGCTCCCCCTACTGTGTCC[A/G]CATCCCCACTCCCAGCCCCTAGAGA
Phenotype
MIM: 611748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTMR11 PubMed Links

Gene Details

Gene
MTMR11
Gene Name
myotubularin related protein 11
There are no transcripts associated with this gene.

Gene
OTUD7B
Gene Name
OTU deubiquitinase 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020205.3 4029 UTR 3 NP_064590.2
XM_011509782.1 4029 UTR 3 XP_011508084.1
XM_011509783.2 4029 UTR 3 XP_011508085.1
XM_011509784.2 4029 UTR 3 XP_011508086.1
XM_011509785.2 4029 UTR 3 XP_011508087.1
XM_011509786.1 4029 UTR 3 XP_011508088.1
XM_011509787.1 4029 UTR 3 XP_011508089.1
XM_011509788.1 4029 UTR 3 XP_011508090.1
XM_017001850.1 4029 UTR 3 XP_016857339.1
XM_017001851.1 4029 UTR 3 XP_016857340.1

View Full Product Details