Product Details

SNP ID
rs3791153
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:151985668 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
ACCAGGATGTGGCATCTCCACCACC[A/G]GGTTTTTGGACCTGAAGGTAGCCAT
Phenotype
MIM: 114085
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 Intron NP_002957.1

View Full Product Details