Product Details

SNP ID
rs4073395
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:11740646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGCATGACCCTTAGGGTATCCTG[C/T]GGATCAGAGGCCCCCCTGGGACTCA
Phenotype
MIM: 608729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGTRAP PubMed Links

Gene Details

Gene
AGTRAP
Gene Name
angiotensin II receptor associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040194.1 Intron NP_001035284.1
NM_001040195.1 Intron NP_001035285.1
NM_001040196.1 Intron NP_001035286.1
NM_001040197.1 Intron NP_001035287.1
NM_020350.4 Intron NP_065083.3
XM_011541799.2 Intron XP_011540101.2
XM_011541800.2 Intron XP_011540102.1
XM_011541802.1 Intron XP_011540104.1

View Full Product Details