Product Details
- SNP ID
-
rs6965245
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:77700426 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGACCCTGTGAAGTATTTCAAGA[C/T]AAAACCAGAGCATTTCACTGAATTT
- Phenotype
-
MIM: 616048
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APTR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs118011229] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APTR
- Gene Name
- Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- RSBN1L
- Gene Name
- round spermatid basic protein 1 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198467.2 |
|
Intron |
|
|
NP_940869.2 |
View Full Product Details