Product Details

SNP ID
rs3087638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133422438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCCTGAATGTGAACCTGAAGT[T/C]CAAAGGACTTGGAAAGCTCTGGAAT
Phenotype
MIM: 610447
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MTG1 PubMed Links

Gene Details

Gene
MTG1
Gene Name
mitochondrial ribosome associated GTPase 1
There are no transcripts associated with this gene.

Gene
SPRN
Gene Name
shadow of prion protein homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012508.3 1401 UTR 3 NP_001012526.2

View Full Product Details