Product Details

SNP ID
rs11216575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117838973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTTACTTGTATGGGGGGAAAAA[A/G]AGGCAGAAGTGAGCTGACAAAACAC
Phenotype
MIM: 606683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXYD6 PubMed Links

Gene Details

Gene
FXYD6
Gene Name
FXYD domain containing ion transport regulator 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164831.2 Intron NP_001158303.1
NM_001164832.2 Intron NP_001158304.1
NM_001164836.2 Intron NP_001158308.1
NM_001164837.2 Intron NP_001158309.1
NM_022003.3 Intron NP_071286.1
Gene
FXYD6-FXYD2
Gene Name
FXYD6-FXYD2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204268.1 Intron NP_001191197.1
NM_001243598.2 Intron NP_001230527.1

View Full Product Details