Product Details
- SNP ID
-
rs4358461
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112717750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAGCTTTGCTGGAAGTGCTTAGA[C/G]AGTTTCTTAGACAACAATGAATATT
- Phenotype
-
MIM: 106410
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANK2
PubMed Links
Gene Details
- Gene
- ANK2
- Gene Name
- ankyrin 2, neuronal
There are no transcripts associated with this gene.
View Full Product Details