Product Details
- SNP ID
-
hCV27049141
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:94868384 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACACCACCTCTTAAATAGGGACA[A/G]TAAGTCCACATCAACACGCCTGCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC442028
PubMed Links
Gene Details
- Gene
- LOC442028
- Gene Name
- uncharacterized LOC442028
There are no transcripts associated with this gene.
- Gene
- TEKT4
- Gene Name
- tektin 4
There are no transcripts associated with this gene.
View Full Product Details