Product Details

SNP ID
rs10945230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70572031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTATAGTAGCCTTACCAAAGCT[G/T]TTACAACACTTTCAGTCTTTTTTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SDHAF4 PubMed Links

Gene Details

Gene
SDHAF4
Gene Name
succinate dehydrogenase complex assembly factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145267.2 Intron NP_660310.2

View Full Product Details